ContactUsLinkComponent
Brady® Alignment Roll for BradyJet™ J7300 Colour Label Printer
  77581-116
New Product
undefined
Brady® Alignment Roll for BradyJet™ J7300 Colour Label Printer
  77581-116
 :  176579
New Product
  • Color:
    White
  • Description:
    Alignment roll
  • Length:
    1524 cm (50 ft.)
  • Cat. No.:
    77581-116
  • Width:
    10.1 cm (4")
Use after replacing the ink cartridge for printhead alignment sequences. For use with the BradyJet™ J7300 color label printer.