Specifications
- Description:LysoBrite™ Lysosome dye, red DND-99
- Size:500 tests
- Cat. No.:MSPP-100-0850
Specifications
About this item
Fluorescent dyes used to label and track lysosomes in live cells - 500 tests.
LysoBrite™ dyes are fluorescent dyes that selectively stain lysosomes and other acidic organelles. Due to their hydrophobic nature, LysoBrite™ dyes permeate live cells and selectively accumulate in the lysosomes via the lysosome’s pH gradient. The fluorescence property of these dyes significantly increases within the lysosome’s acidic environment; this fluorescence can be measured using flow cytometry and fluorescence imaging. Due to their selective fluorescence inside lysosomes, LysoBrite™ dyes provide low background and high contrast during imaging. They can be used with proliferating and non-proliferating cells, and are suitable for suspension and adherent cells.
CAS No: 67-68-5