Caspase-3/7 Activity Flow Cytometry Kit, Green
For detection of active caspase-3 in mammalian cells undergoing apoptosis - 100 tests.
- TF2-DEVD-FMK detects caspase-3/7 activity
- Excitation wavelength: 503 nm
- Emission wavelength: 525 nm
- Extinction coefficient: 75,000 cm⁻¹M⁻¹
- Easily scalable for high-throughput assays
- Verified for use with flow cytometry
Efficiently detect active caspase-3 in mammalian cells undergoing apoptosis by using the Caspase-³/₇ activity flow cytometry kit, green. This reagent kit uses a quick and simple staining protocol that is highly sensitive and compatible with standard flow cytometers.
Caspase-3 and caspase-7 have a proteolytic substrate selectivity for the peptide sequence Asp-Glu-Val-Asp (DEVD). This kit includes TF2-DEVD-FMK a cell-permeable and non-toxic fluorescent substrate as well as an assay buffer and propidium iodide. When added to apoptotic cells, TF2-DEVD-FMK irreversibly binds to activated caspase-3, resulting in the retention of the green fluorescent reagent within cells that can be detected by flow cytometry.
Caspases are a family of proteases that play a central role in cellular apoptosis through the cleavage of select proteins, and result in the disassembly of the cell (Thornberry & Lazebnik Science, 1998). This process is switched on by pro-apoptotic signals from death receptors or stimuli such as cytotoxic reagents, which activate the initiator caspases and lead to activation of the effector caspases, caspase-3 and caspase-7 (Thornberry & Lazebnik Science, 1998). Caspase-3/7 Activity Flow Cytometry Kit, Green is useful for screening caspase-3 inhibitors and quantifying apoptotic cells containing active caspase-3.
CAS No: 67-68-5
: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
- Description:Caspase-3/7 activity flow cytometry kit, green
- Size:100 tests
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



