Specifications
- Description:MegaCult™-C Staining kit for CFU-Mk
- Size:1 kit
- Environmentally Preferable:
- Cat. No.:MSPP-04962
Specifications
About this item
Staining kit for human CFU-Mk assays - 1 kit.
Stain fixed and dehydrated CFU-Mk colonies cultured in MegaCult™-C collagen-based medium using the reagents included in MegaCult™-C staining kit for CFU-Mk.
Megakaryocytes and platelets expressing the glycoprotein IIb/IIIa are stained with anti-human CD41 antibody and an alkaline phosphatase detection system. The kit is also suitable for the detection of bipotential erythroid-megakaryocyte progenitor cells (BFU-E/Mk).