About this item
Amine-labeling fluorescent dye for live/dead staining of mammalian cells - 100 tests.
GloCell™ Fixable viability dye violet 450 is a fluorescent cell viability dye for staining live/dead mammalian cells in applications such as flow cytometry. The dye irreversibly binds intracellular and cell surface amine groups and can be utilized prior to fixation, permeabilization, or cryopreservation. Cells with compromised plasma membranes become permeable to the GloCell™ dye, resulting in a greater fluorescence compared to live cells in a cell viability assay.
GloCell™ Fixable Viability Dye Violet 450 is excited by the violet laser at 405 nm and it has a fluorescence emission maximum of 450 nm that can be detected using the 440/40 or 440/50 band pass filter used to detect Pacific Blue™.
CAS No: 67-68-5