Order Entry
United States
ContactUsLinkComponent
1,1,2,2-Tetrachloroethane-[D₂] 99.5+ atom % D for NMR spectroscopy
1,1,2,2-Tetrachloroethane-[D₂] 99.5+ atom % D for NMR spectroscopy
Catalog # TS464110100
New Product
CAS Number:  
1,1,2,2-Tetrachloroethane-[D₂] 99.5+ atom % D for NMR spectroscopy
Catalog # TS464110100
Supplier Number:  464110100
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    10 ml
  • Cat. No.:
    TS464110100

Specifications

About this item

 :
 :169.83 g/mol
UN:1702
CAS Number:33685-54-0
Formula:Cl₂CDCDCl₂
MDL Number:MFCD00037672