Diethylene glycol monomethyl ether 99%
Supplier: Thermo Scientific Chemicals
New Product
Certificates
About this item
Synonyms: 2-(2-Methoxyethoxy)ethanol, Diethylene glycol methyl ether
CAS Number:111-77-3
:
Formula:C₅H₁₂O₃
MDL Number:MFCD00002871
Boiling Pt:193
Melting Pt:-65
Density:1.01 g/cm³
Molecular Weight:120.15 g/mol
Flash Pt:83 °C
Merck Index:12,06116
EINECS:203-906-6
:GHS08
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4250 to 1.4270 (20°C, 589 nm)
- Water:≤0.2 %
- GC:≥98.5 %
- Color scale:≤15 APHA
Specification Test Results
Frequently Bought Together