Order Entry
United States
ContactUsLinkComponent
Methyl palmitoleate 99% analytical standard for GC
Methyl palmitoleate 99% analytical standard for GC
Catalog # TS460400010
New Product
CAS Number:  
Methyl palmitoleate 99% analytical standard for GC
Catalog # TS460400010
Supplier Number:  460400010
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 g
  • Cat. No.:
    TS460400010

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4500 to 1.4520 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

About this item

 :
 :268.44 g/mol
Melting Pt:.5 °C
Density:0.88 g/cm³
Storage Temperature:Freezer
CAS Number:1120-25-8
Formula:C₁₇H₃₂O₂
MDL Number:MFCD00042911