1,4-Dioxane, extra dry 99.8% stabilized, AcroSeal®
Catalog # TS450171000
Supplier: Thermo Scientific Chemicals
New Product
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Size:100 ml
- Cat. No.:TS450171000
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 %
- GC:≥99.75 %
- Stabilizer:2 to 5 ppm BHT
- Residue after evaporation:≤0.001 %
- Acidity (CH3COOH):≤0.002
- Peroxides:≤0.005 % (as H2O2)
Specification Test Results
About this item
:
:88.11 g/mol
Boiling Pt:101 °C (760 mmHg)
Melting Pt:12 °C
Density:1.034 g/cm³
Flash Pt:12 °C
UN:1165
ADR:3,II
:GHS02
:GHS07
:GHS08
Storage Temperature:Ambient
CAS Number:123-91-1
Formula:C₄H₈O₂
MDL Number:MFCD00006571