

Dichloromethane, extra dry 99.9% stabilized, AcroSeal®
Catalog # TS448371000
Supplier: Thermo Scientific Chemicals
New Product
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Size:4x 25 ml
- Cat. No.:TS448371000
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Water:≤0.003 % (Coulometric)
- GC:≥99.85 %
- Stabilizer:Approx. 50 ppm Amylene
- Residue after evaporation:≤0.0005 %
- Filt.over 0.2 micron PFTE:Filtered
Specification Test Results
About this item
:
:84.93 g/mol
Boiling Pt:39…40 °C (760 mmHg)
Melting Pt:–97 °C
Density:1.325 g/cm³
UN:1593
ADR:6.1,III
:GHS07
:GHS08
Storage Temperature:Ambient
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881