ContactUsLinkComponent
Human Pluripotent Stem Cell Naïve State qPCR Array
Human Pluripotent Stem Cell Naïve State qPCR Array
  MSPP-07521
 :  
undefined
Human Pluripotent Stem Cell Naïve State qPCR Array
  MSPP-07521
 :  07521
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Human pluripotent stem cell Naïve state qPCR array
  • Size:
    1 unit
  • Cat. No.:
    MSPP-07521
For characterization of gene expression associated with hPSC naïve or primed states.
Characterize your hPSCs by accurately measuring their gene expression profile to determine their status along the spectrum from naïve to primed pluripotency. qPCR is used to determine changes in steady-state mRNA levels of gene expression across multiple samples, generally normalized to the relative expression of internal control genes. Gene-specific primers amplify target sequences within cDNA pools reverse-transcribed from mRNA and, as with TaqMan® technology, hybridized sequence-specific probes provide a fluorescent signal from the 5' exonuclease activity of Taq DNA polymerase. The accumulation rate of the fluorescent signal is used to quantify the sample cDNA, and thereby the amount of mRNA in the original cell lysate.
The Human Pluripotent Stem Cell (hPSC) Naïve State qPCR Array has validated primers and probes to detect 90 genes preselected for their demonstrated differential expression in naïve state and primed hPSCs and their derivatives, as well as 6 endogenous control genes. TBP (TATA box-binding protein) qPCR Control Template is provided as a synthetic DNA positive control. Naïve state hPSCs are self-renewing, retain the properties of the pre-implantation blastocyst, and can differentiate to both embryonic and extra-embryonic lineages. Primed hPSCs are also self-renewing, but they are restricted to differentiation along the three embryonic germ layers. Additional gene information and a qPCR analysis tool are available here.
 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').