DNA Loading Dye
Catalog # MSPP-7918
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- Description:DNA loading dye
- Size:3 ml
- Cat. No.:MSPP-7918
Specifications
About this item
Ready-to-use marker dye for loading DNA samples into agarose gel wells and tracking migration during electrophoresis.
DNA Loading Dye is a ready-to-use marker dye containing orange G (0.4%), bromophenol blue (0.03%), and xylene cyanol FF (0.03%), in 15% Ficoll® 400, 10 mM Tris-HCl (pH 7.5), and 50 mM EDTA (pH 8.0). The dye is used for loading DNA samples into agarose gel wells and tracking migration during electrophoresis. In a 0.5 to 1.4% agarose gel in 0.5X TBE, xylene cyanol FF migrates at approximately 4 kb, bromophenol blue at approximately 300 bp, and orange G at approximately 50 bp. This product is free of nuclease activity.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').