Order Entry
United States
ContactUsLinkComponent
EasySep™ Total Nucleic Acid Extraction Kit
EasySep™ Total Nucleic Acid Extraction Kit
Catalog # MSPP-100-1079
CAS Number:  
undefined
EasySep™ Total Nucleic Acid Extraction Kit
Catalog # MSPP-100-1079
Supplier Number:  100-1079
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
  • Method / Format:
  • Refine target molecule:
  • Sample type:
  • Target molecule:
  • Description:
    EasySep™ Total nucleic acid extraction kit, Genome editing: Nucleic acid purification
  • Size:
    100 preparations
  • Environmentally Preferable:
  • Cat. No.:
    MSPP-100-1079

Specifications

About this item

Immunomagnetic extraction of total nucleic acid (DNA and RNA).

  • Scale up nucleic acid extractions of up to 16 or 96 samples
  • Save sample processing time by avoiding spinning columns and centrifugation steps
  • Simplify your workflow with an easy-to-use system that eliminates the use of toxic reagents
  • Achieve consistent extraction of nucleic acids from a variety of sample types such as whole blood and cell suspensions

Efficiently extract total nucleic acids (DNA and RNA) from whole blood, cell suspensions (e.g. PBMCs, hPSCs, cultured cells), and EasySep™-isolated cells. Designed with easy-to-use and scalable protocols, the EasySep™ Total Nucleic Acid Extraction Kit uses magnetic bead technology that allows you to avoid the use of spin -columns or toxic reagents. Nucleic acids extracted with this kit are highly pure and can be used in downstream applications such as qPCR.Use this kit to magnetically label total nucleic acids in your sample with EasySep™ Total Nucleic Acid RapidSpheres™. Then, process the samples—without the use of columns—using a magnet (sold separately). Pipette off the unlabeled, undesired components before removing the sample from the magnet and recovering your labeled nucleic acids. Protocols are available for extracting nucleic acid using 1.7 ml microcentrifuge tubes and the ErythroClear™ Magnet (Catalog MSPP-01737). To scale up your extractions, process samples in 96-well PCR plates with a 96-Well PCR Microplate Magnet (Catalog #MSPP-100-1304).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').