To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Application:
- Method / Format:
- Refine target molecule:
- Sample type:
- Target molecule:
- Description:Total RNA purification kit
- Size:50 preparations
- Environmentally Preferable:
- Cat. No.:MSPP-79040
For purification of RNA from cells.
The Total RNA Purification Kit uses a membrane-based system to quickly purify intact total RNA from 1×10² to 5×10⁶ cells without an organic extraction step, eliminating the need for additional purification steps. The Total RNA Purification Kit includes an on-column DNase I treatment to significantly reduce genomic DNA contamination and ensure the purified RNA is suitable for sensitive downstream applications. Intact total RNA can be isolated in as little as 30 minutes. The Total RNA Purification Kit is not recommended for the purification of small RNAs (<200 nt), such as miRNA.