Human recombinant CTLA-4-Ig
Catalog # 77578-792
Supplier: Bio X Cell
New Product
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Species:Human
- Size:1 mg
- Protein/Peptide Name:CTLA-4-Ig
- Cat. No.:77578-792
Specifications
About this item
Fusion Protein