
Trypan blue 0.4% in PBS
Catalog # MSPP-70-50
Supplier: Stemcell Technologies
CAS Number:
Specifications
- Size:100 ml
- Cat. No.:MSPP-70-50
Specifications
About this item
:
:960.82 g/mol
:GHS08
Storage Temperature:Ambient
CAS Number:72-57-1
Formula:C₃₄H₂₄N₆Na₄O₁₄S₄
MDL Number:MFCD00003969