Buccutite™ Rapid Oligo Antibody Conjugation Kit
77462-894
:
- Size:2
- Cat. No.:77462-894
- Includes:2 labellings
optimized for labelling 100 µg protein. Buccutite™ rapid oligo antibody conjugation kit is optimized for conjugating 100 µg protein with an amino-modified oligo.
- Robust protocol
- Consistent results
- 12-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
: Shipped at RT.
: Research use only.