ContactUsLinkComponent
Buccutite™ Rapid Crosslinked APC Antibody Labelling Kit
Buccutite™ Rapid Crosslinked APC Antibody Labelling Kit
Catalog # 77462-878
Supplier:  AAT BIOQUEST INC
CAS Number:  
undefined
Buccutite™ Rapid Crosslinked APC Antibody Labelling Kit
Catalog # 77462-878
Supplier:  AAT BIOQUEST INC
Supplier Number:  5406
CAS Number:  
Specifications
  • Size:
    2
  • Cat. No.:
    77462-878
  • Includes:
    2 labellings
Specifications
About this item
Production scale optimized for labelling 1 mg antibody per reaction. Buccutite™ Rapid crosslinked APC antibody labelling kits, designed for large-scale production, offer a convenient and efficient method to label antibodies with APC, PE and iFluor® tandem dyes.
  • Robust protocol
  • Consistent results
  • Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Delivery: Shipped with blue ice.
Caution: Research use only.