Live or Dead™ Fixable Dead Cell Staining Kit
Catalog # 77461-912
Supplier: AAT BIOQUEST INC
CAS Number:
Specifications
- Size:200 tests
- Cat. No.:77461-912
Specifications
About this item
IR Fluorescence. Our Live or Dead™ fixable dead cell staining kits are a set of tools for labelling cells for fluorescence microscopic investigations of cellular functions.
- The kit provides all the essential components with an optimized cell-labelling protocol
- The fluorescent dye used in the kit is well excited with the IR laser (808 nm) and emits fluorescence at 878 nm (885/40 bandpass filter).
This particular kit is designed to uniformly label fixed mammalian cells in infrared fluorescence for flow cytometry applications with IR laser excitation.
Delivery: Shipped at RT.
Caution: Research use only.