ContactUsLinkComponent
Amplite® Fluorimetric Sodium Ion Quantification Kit
Amplite® Fluorimetric Sodium Ion Quantification Kit
Catalog # 77461-868
Supplier:  AAT BIOQUEST INC
CAS Number:  
undefined
Amplite® Fluorimetric Sodium Ion Quantification Kit
Catalog # 77461-868
Supplier:  AAT BIOQUEST INC
Supplier Number:  21325
CAS Number:  
Specifications
  • Size:
    100 Tests
  • Cat. No.:
    77461-868
  • No. of tests:
    100 tests
Specifications
About this item
Designed for consistent and dependable results. Amplite® fluorimetric sodium ion quantification kit uses our robust sodium ion indicator dye, SoNa™ 520, which exhibit great fluorescence intensity enhancement upon binding to sodium ions.
  • Robust protocol
  • SoNa™ 520 is perhaps the most robust sodium ion indicator with high selectivity
  • 12 month shelf life
This microplate-based assay kit requires extremely small amount of sample, it is particularly suitable for the determination of sodium ion concentration in microvolume format.
Delivery: Shipped at RT.
Caution: Research use only.