Order Entry
United States
ContactUsLinkComponent
 
Anti-IgG Goat Polyclonal Antibody *Cross Adsorbed* (iFluor® 680)
 
undefined
Anti-IgG Goat Polyclonal Antibody *Cross Adsorbed* (iFluor® 680)

 

  • Catalog No:
  • 77462-530
  • 77462-528
  • Antigen Symbol:
  • IgG
  • IgG
  • Antigen Name:
  • Immunoglobulin G
  • Immunoglobulin G
  • Antibody Type:
  • Secondary
  • Secondary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • iFluor® 680
  • iFluor® 680
  • Reactivity:
  • Human
  • Human
  • Host:
  • Goat
  • Goat
  • Isotype:
  • IgG (H, L)
  • IgG (H, L)
  • Western Blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Flow Cytometry:
  • Yes
  • Yes
  • Cross Adsorption:
  • Minimal cross reaction to human, horse, mouse, rabbit and bovine
  • Minimal cross reaction to human, horse, mouse, rabbit and bovine
  • Storage Temperature:
  • Freeze (<–15 °C); minimize light exposure
  • Freeze (<–15 °C); minimize light exposure
  • Shipping Temperature:
  • 20
  • 20

 

Frequently Bought Together