Specifications
- Frame color:White
- No. of wells:96
- Profile:Low
- Skirt:Semi-Skirted
- Well Color:White
- Description:Cy3.5 Dye qPCR calibration plate
- Environmentally Preferable:
- Cat. No.:77463-000
Specifications
About this item
The Cy3.5 dye qPCR calibration plate can be used to maintain the 7500 real-time PCR system with a fast 96-well block.
- The qPCR calibration plate might significantly improve qPCR results with multiplexing by more accurately representing fluorescent spectra used in your real-time experiments
- This calibration plate is ready to use without any additional preparation steps required
For most qPCR instruments, the necessary calibrations should be run at least every six months.