ContactUsLinkComponent
VIC Dye qPCR Calibration Plate *Optimized for ABI7500 Fast 96-Well*
VIC Dye qPCR Calibration Plate *Optimized for ABI7500 Fast 96-Well*
Catalog # 77462-988
Supplier:  AAT BIOQUEST INC
undefined
VIC Dye qPCR Calibration Plate *Optimized for ABI7500 Fast 96-Well*
Catalog # 77462-988
Supplier:  AAT BIOQUEST INC
Supplier Number:  67002
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Frame color:
    White
  • No. of wells:
    96
  • Profile:
    Low
  • Skirt:
    Semi-Skirted
  • Well Color:
    White
  • Description:
    VIC Dye qPCR calibration plate
  • Environmentally Preferable:
  • Cat. No.:
    77462-988
Specifications
About this item
The VIC dye qPCR calibration plate can be used to maintain the 7500 real-time PCR system with fast 96-well block.
  • The qPCR calibration plate might significantly improve qPCR results with multiplexing by more accurately representing fluorescent spectra used in your real-time experiments
  • This calibration plate is ready to use without any additional preparation steps required
For most qPCR instruments, the necessary calibrations should be run at least every six months.
Caution: Avoid freeze-thawing plates. It is not recommended to use the plates more than three times. Plates can be stored for up to 12 months. Freeze (<−15 °C), and minimize light exposure.