ContactUsLinkComponent
Cleanroom Arc Flash Booties
Cleanroom Arc Flash Booties
Supplier:  Sellstrom
New Product
Certificates
undefined
Cleanroom Arc Flash Booties
Supplier:  Sellstrom
New Product
Specifications
  • Catalog No:
  • 77575-486
  • 77575-518
  • 77575-496
  • Model:
  • CR-BOOTIE
  • CR-BOOTIE
  • CR-BOOTIE
  • Garment application:
  • Industrial, Electronics/semi-conductor
  • Industrial, Electronics/semi-conductor
  • Industrial, Electronics/semi-conductor
  • Fabric:
  • Nomex
  • Nomex
  • Nomex
  • Weight:
  • 1.25 lbs.
  • 1.25 lbs.
  • 1.25 lbs.