Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
iFluor®
iFluor® 440, Styramide *Superior Replacement for Alexa Fluor 440 tyramide*
iFluor® 440, Styramide *Superior Replacement for Alexa Fluor 440 tyramide*
iFluor® 440, Styramide *Superior Replacement for Alexa Fluor 440 tyramide*
Catalog # 77462-262
Supplier:
AAT Bioquest
CAS Number:
Expand image
iFluor® 440, Styramide *Superior Replacement for Alexa Fluor 440 tyramide*
Catalog # 77462-262
Supplier:
AAT Bioquest
Supplier Number:
44900
CAS Number:
Specifications
Size:
100 slides
Cat. No.:
77462-262
Specifications
About this item
Product Details
:
Product Details