Orders LinkContactUsLinkComponent
Funnels, PTFE
Funnels, PTFE
Funnels, PTFE
Certificates
5 Options Available Below
Product Details & Documents
The PTFE Funnels are ideal for general laboratory use as it has good resistance to chemical attack.
  • Lightweight plastic funnels can be easily cleaned after use
  • Five handy sizes from 30 to 150 mm
  • Crevice free interior
  • Heavy wall construction
Virgin PTFE that conforms to FDA and USP Class VI requirements is used in the manufacture of these funnels.
Certifications: FDA and EC guidelines, BSE/TSE free.
Product Family Options
Product Information
  • Material
    Height
    Stem diameter
    Top diameter
    Availability
    Price
  • PTFE
    49 mm
    8 mm
    30 mm
    Log in for availability
    Log in for availability
    Each
    Each - $28.30
    $28.30
    Each
       $28.30
    Supplier Number: A105-30
  • PTFE
    82 mm
    10 mm
    50 mm
    Log in for availability
    Log in for availability
    Each
    Each - $38.50
    $38.50
    Each
       $38.50
    Supplier Number: A105-50
  • PTFE
    110 mm
    12 mm
    70 mm
    Log in for availability
    Log in for availability
    Each
    Each - $56.30
    $56.30
    Each
       $56.30
    Supplier Number: A105-70
  • PTFE
    140 mm
    14 mm
    100 mm
    Log in for availability
    Log in for availability
    Each
    Each - $116.00
    $116.00
    Each
       $116.00
    Supplier Number: A105-100
  • PTFE
    195 mm
    18 mm
    150 mm
    Log in for availability
    Log in for availability
    Each
    Each - $202.00
    $202.00
    Each
       $202.00
    Supplier Number: A105-150