Order Entry
United States
ContactUsLinkComponent
1,18-Octadecanediol ≥98.0% (by GC)
1,18-Octadecanediol ≥98.0% (by GC)
Catalog # TCO0563-250MG
Supplier:  TCI America
New Product
CAS Number:  
1,18-Octadecanediol ≥98.0% (by GC)
Catalog # TCO0563-250MG
Supplier:  TCI America
Supplier Number:  O0563-250MG
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    250 mg
  • Cat. No.:
    TCO0563-250MG

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    97.0 to 101.0 deg-C

Specification Test Results

About this item

 :
 :286.50 g/mol
Boiling Pt:211 °C
CAS Number:3155-43-9
Formula:C₁₈H₃₈O₂
MDL Number:MFCD17168570