Orders LinkContactUsLinkComponent
Vivaflow® Peristaltic Pump Head
Vivaflow® Peristaltic Pump Head
Supplier:   Sartorius
Vivaflow® Peristaltic Pump Head
Supplier:  Sartorius
Certificates
Vivaflow® Peristaltic Pump Head
Vivaflow® Peristaltic Pump Head
2 Options Available Below
Product Details & Documents
Recommended for processing samples with Vivaflow® tangential flow filtration (TFF) cassettes, these Watson-Marlow pumps are ideal for ultrafiltration and diafiltration applications.
  • Simple and intuitive set-up
  • Precise flow control
  • Avoid contamination
  • Low maintenance
Peristaltic pump heads offer the flexibility, performance, and control needed for other filtration and fluid transfer applications. Pump head VF-APH0001-1 is suitable for most Vivaflow® configurations. To operate two Vivaflow® 200 cassettes in parallel with a Y-connector, use pump head VF-APH0002-1.
Product Family Options
Product Information
  • Availability
    Price
  • 77590-228
    Peristaltic pump head for 1.6 mm W.T. tubing
    Peristaltic pump head for 1.6 mm W.T. tubing
    Log in for availability
    Log in for availability
    Each
    Each - $787.54
    $787.54
    Each
       $787.54
    Supplier Number: VF-APH0001-1
  • 77590-230
    Peristaltic pump head for 2.4 mm W.T. tubing
    Peristaltic pump head for 2.4 mm W.T. tubing
    Log in for availability
    Log in for availability
    Each
    Each - $787.54
    $787.54
    Each
       $787.54
    Supplier Number: VF-APH0002-1