DNase I-XT Reaction Buffer
77575-352
New Product
:
- Concentration:10X
- Description:DNase I-XT reaction buffer
- Size:6 ml
- Cat. No.:77575-352
New England Biolabs supplies a 10X reaction buffer with all of its enzymes.
- For use with DNase I-XT (M0570)
- Supplied in easy to use 10X format
At a 10X concentration this reaction buffer assures optimal activity of the enzyme.