Human Recombinant LCN2 (from HEK293 cells)
Catalog # 77540-120
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:50 µg
- Storage Conditions:4 °C
- Endotoxin Content:<1.0 EU/µg of the protein as determined by the LAL method
- Biological Activity:Measured by its ability to bind Iron(III) dihydroxybenzoic acid [Fe(DHBA)3]. The binding of Fe(DHBA)3 results in the quenching of Trp fluorescence in recombinant human Lipocalin-2. Recombinant human Lipocalin-2 can bind >1.5 μM of Fe(DHBA)3.
- Reconstitution Instructions:It is recommended that sterile water be added to the vial to prepare a stock solution of 0.2 µg/µl. Centrifuge the vial at 4 °C before opening to recover the entire contents.
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:lipocalin 2
- UniProtKB:P80188
- Protein/Peptide Name:LCN2
- Purity:>97% as determined by SDS-PAGE
- Molecular Weight:The secreted recombinant human LCN2 consists of 195 amino acids and predicts a molecular mass of 22.7 kDa. As a result of glycosylation, it migrates as an approximately 25.7 kDa protein in SDS-PAGE under reducing conditions.
- Endotoxin Level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Cat. No.:77540-120