Order Entry
United States
ContactUsLinkComponent
Human Recombinant IL11 (from E. coli)
Human Recombinant IL11 (from E. coli)
Supplier:  Creative Biomart
New Product
Certificates
undefined
Human Recombinant IL11 (from E. coli)
Supplier:  Creative Biomart
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    4 °C
  • Endotoxin Content:
    <1 EU/µg of rHuIL-11 as determined by LAL method
  • Biological Activity:
    Fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using murine B9-11 cells is less than 1 ng/ml, corresponding to a specific activity of >1.0×10⁶ IU/mg.
  • Reconstitution Instructions:
    Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1 - 1.0 mg/ml. Stock solutions should be apportioned into working aliquots and stored at ≤−20 °C. Further dilutions should be made in appropriate buffered solutions.
  • Endotoxin-free:
    No
  • Carrier-Free:
    No
  • Protease-free:
    No
  • Animal-Free:
    No
  • Protein Synonyms:
    interleukin 11
  • UniProtKB:
    P20809
  • Protein/Peptide Name:
    IL11
  • Purity:
    >95% by SDS-PAGE and HPLC analyses
  • Molecular Weight:
    Approximately 19.1 kDa, a single non-glycosylated polypeptide chain containing 179 amino acids
  • Sequence:
    MPGPPPGPPRVSPDPRAELDSTVLLTRSLLADTRQLAAQLRDKFPADGDHNLDSLPTLAMSAGALGALQLPGVLTRLRADLLSYLRHVQWLRRAGGSSLKTLEPELGTLQARLDRLLRRLQLLMSRLALPQPPPDPPAPPLAPPSSAWGGIRAAHAILGGLHLTLDWAVRGLLLLKTRL
  • Endotoxin Level:
    Low
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping Temperature:
    20

Specifications

Frequently Bought Together