Human Recombinant HBv (from E. coli)
Supplier: Creative Biomart
New Product
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:4 °C
- Endotoxin Content:<10 EU/mg as determined by LAL method
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:Hepatitis B Virus
- Protein/Peptide Name:HBv
- Purity:≥95% as determined by SDS-PAGE
- Endotoxin Level:Low
- Concentration:0.2 mg/ml by BCA
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Tested Applications:HBV Core antigen is stable in ELISA and western blots, excellent antigen for detection of HBV with minimal specificity problems
Specifications
Frequently Bought Together