Human Recombinant GBA2 (from HEK293 cells)
Catalog # 77539-574
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:20 µg
- Storage Conditions:4 °C
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:glucosidase, beta (bile acid) 2
- UniProtKB:Q9HCG7
- Protein/Peptide Name:GBA2
- Purity:>80% as determined by SDS-PAGE and Coomassie blue staining
- Molecular Weight:104.5 kDa
- Endotoxin Level:Low
- Concentration:>50 µg/ml as determined by microplate BCA method
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Cat. No.:77539-574