Human Recombinant FSHR (from HEK293 cells)
Catalog # 77539-546
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:20 µg
- Storage Conditions:4 °C
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:follicle stimulating hormone receptor
- UniProtKB:P23945
- Protein/Peptide Name:FSHR
- Purity:>85% as determined by SDS-PAGE
- Molecular Weight:43.5 kDa
- Sequence:CHHRICHCSNRVFLCQESKVTEIPSDLPRNAIELRFVLTKLRVIQKGAFSGFGDLEKIEISQNDVLEVIEADVFSNLPKLHEIRIEKANNLLYINPEAFQNLPNLQYLLISNTGIKHLPDVHKIHSLQKVLLDIQDNINIHTIERNSFVGLSFESVILWLNKNGIQEIHNCAFNGTQLDELNLSDNNNLEELPNDVFHGASGPVILDISRTRIHSLPSYGLENLKKLRARSTYNLKKLPTLEKLVALMEASLTYPSHCCAFANWRRQISELHPICNKSILRQEVDYMTQARGQRSSLAEDNESSYSRGFDMTYTEFDYDLCNEVVDVTCSPKPDAFNPCEDIMGYNILR
- Endotoxin Level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Cat. No.:77539-546