Bovine Native FG (from Serum)
Catalog # 77539-460
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Native
- Source:Serum
- Species:Bovine
- Size:1 g
- Storage Conditions:4 °C
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:fibrinogen
- Protein/Peptide Name:FG
- Purity:>95% (SDS-PAGE)
- Endotoxin Level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Tested Applications:Western blotting, ELISA, etc.,
- Cat. No.:77539-460
Specifications
About this item
Purified from pooled 'Normal bovine serum'.