Human Recombinant ERBB3 (from Insect cells)
Catalog # 77539-414
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Size:20 µg
- Storage Conditions:4 °C
- Endotoxin Content:<1.0 EU/µg of the protein as determined by the LAL method
- Biological Activity:No Kinase Activity. Using the Octet RED System, the affinity constant (Kd) of human ErBB3-GST bound to human NRG1 (aa 2-224) was 20 nM.
- Reconstitution Instructions:It is recommended that sterile water be added to the vial to prepare a stock solution of 0.2 µg/µl. Centrifuge the vial at 4 °C before opening to recover the entire contents.
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:ERBB3 v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian)
- UniProtKB:P21860
- Protein/Peptide Name:ERBB3
- Purity:>75% as determined by SDS-PAGE
- Molecular Weight:The recombinant human ErbB3 /GST chimera consists of 573 amino acids and has a calculated molecular mass of 65 KDa. It migrates as an approximately 65 KDa band as predicted in SDS-PAGE under reducing conditions.
- Endotoxin Level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Cat. No.:77539-414