Human Recombinant CTLA4 (from HEK293 cells)
Catalog # 77539-206
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:100 µg
- Storage Conditions:4 °C
- Endotoxin Content:<1.0 EU/µg by the LAL method
- Reconstitution Instructions:It is recommended that sterile water be added to the vial to prepare a stock solution of 0.2 µg/µl. Centrifuge the vial at 4 °C before opening to recover the entire contents.
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:Ctla4 cytotoxic T-lymphocyte-associated protein 4
- UniProtKB:P16410
- Protein/Peptide Name:CTLA4
- Purity:>95% as determined by SDS-PAGE
- Molecular Weight:The protein has a calculated MW of 15.4 kDa. The protein migrates as 25 - 30 kDa under reducing (R) condition (SDS-PAGE) due to different extent glycosylation.
- Endotoxin Level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Cat. No.:77539-206