Bovine SOD1 (from Kidney tissue)
Supplier: Creative Biomart
New Product
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Source:Kidney tissue
- Species:Bovine
- Storage Conditions:4 °C
- Biological Activity:3000 U/mg Protein. The method of assay is based on the ability of superoxide dismutase to compete with ferricytochrome C for superoxide anions generated by the xanthine oxidase system. This results in inhibition of the rate of reduction of ferricytochrome C.
- Reconstitution Instructions:Soluble in distilled water or dilute buffer
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:superoxide dismutase 1, soluble
- UniProtKB:P00441
- Protein/Peptide Name:SOD1
- Purity:95% (biuret)
- Endotoxin Level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
Specifications
Frequently Bought Together