E. coli Recombinant CdtB (from E. coli)
Catalog # 77539-032
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:E. coli
- Size:25 µg
- Storage Conditions:4 °C
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:Cytolethal distending toxin B
- UniProtKB:A5LH53
- Protein/Peptide Name:CdtB
- Purity:>90% by SDS-PAGE
- Endotoxin Level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Cat. No.:77539-032