Human Recombinant CD22 (from HEK293 cells), Alexa Fluor® 647
Supplier: Creative Biomart
New Product
Certificates
About this item
Specifications
- Conjugation:Alexa Fluor® 647
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:4 °C
- Endotoxin Content:<1.0 EU/ μg by the LAL method
- Biological Activity:Immobilized Human CD22 Protein, His-tagged at 2 μg/ml (100 μl/well) can bind Anti-Human CD22 MAb, Human IgG1 with a linear range of 0.1 - 2 ng/ml (QC tested).
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:CD22 molecule
- UniProtKB:P20273
- Protein/Peptide Name:CD22
- Purity:>95% as determined by SDS-PAGE
- Molecular Weight:The protein has a calculated MW of 57.8 kDa. The protein migrates as 90 - 100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
- Endotoxin Level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
Specifications
Frequently Bought Together