Order Entry
United States
ContactUsLinkComponent
 
Human Recombinant BTNL2 (from CHO cells)
 
undefined
Human Recombinant BTNL2 (from CHO cells)

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Storage Conditions:
    4 °C
  • Endotoxin Content:
    <0.06 EU/µg as determined by lal
  • Biological Activity:
    A butyrophilin-like molecule that functions to inhibit T cell activation
  • Reconstitution Instructions:
    Reconstitute at 100 μg/ml in sterile PBS
  • Endotoxin-free:
    No
  • Carrier-Free:
    No
  • Protease-free:
    No
  • Animal-Free:
    No
  • Protein Synonyms:
    butyrophilin-like 2 (MHC class II associated)
  • UniProtKB:
    Q9UIR0
  • Protein/Peptide Name:
    BTNL2
  • Purity:
    ≥98% by SDS-PAGE under reducing conditions
  • Molecular Weight:
    74 KDa (monomer)
  • Endotoxin Level:
    Low
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping Temperature:
    20

 

Frequently Bought Together