Human Recombinant ACVR2B (from CHO cells)
Catalog # 77538-556
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:CHO cells
- Species:Human
- Size:100 µg
- Storage Conditions:4 °C
- Endotoxin Content:<0.1 EU/µg of the protein
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:activin A receptor, type IIB
- UniProtKB:Q13705
- Protein/Peptide Name:ACVR2B
- Purity:>96%
- Endotoxin Level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Cat. No.:77538-556