Order Entry
United States
ContactUsLinkComponent
Human Recombinant ACE2 (from Mammalian cells)
Human Recombinant ACE2 (from Mammalian cells)
Supplier:  Creative Biomart
New Product
Certificates
undefined
Human Recombinant ACE2 (from Mammalian cells)
Supplier:  Creative Biomart
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Mammalian cells
  • Species:
    Human
  • Storage Conditions:
    4 °C
  • Endotoxin Content:
    <1 EU/μg by the LAL method
  • Reconstitution Instructions:
    It is recommended that sterile water be added to the vial to prepare a stock solution of 0.25 mg/ml. Centrifuge the vial at 4 °C before opening to recover the entire contents.
  • Endotoxin-free:
    No
  • Carrier-Free:
    No
  • Protease-free:
    No
  • Animal-Free:
    No
  • Protein Synonyms:
    angiotensin I converting enzyme 2
  • UniProtKB:
    Q9BYF1
  • Protein/Peptide Name:
    ACE2
  • Purity:
    >95% as determined by Bis-Tris PAGE. >95% as determined by HPLC.
  • Molecular Weight:
    The protein has a predicted MW of 86.5 kDa. Due to glycosylation, the protein migrates to 95 - 110 kDa based on Bis-Tris PAGE result.
  • Endotoxin Level:
    Low
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping Temperature:
    20

Specifications

Frequently Bought Together