Order Entry
United States
ContactUsLinkComponent
Human Recombinant HSP90AA1 (from E. coli)
Human Recombinant HSP90AA1 (from E. coli)
Supplier:  Creative Biomart
Certificates
Human Recombinant HSP90AA1 (from E. coli)
Supplier:  Creative Biomart

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    4 °C
  • Endotoxin-free:
    No
  • Carrier-Free:
    No
  • Protease-free:
    No
  • Animal-Free:
    No
  • Protein Synonyms:
    heat shock protein 90kDa alpha (cytosolic), class A member 1
  • UniProtKB:
    Q86SX1
  • Protein/Peptide Name:
    HSP90AA1
  • Purity:
    >95% as determined by: Analysis by RP-HPLC and by analysis by reducing andnon-reducing SDS-PAGE coomassie
  • Endotoxin Level:
    Low
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping Temperature:
    20

Specifications

Frequently Bought Together