Human Recombinant GNB1 (from E. coli)
Supplier: Creative Biomart
About this item
3 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 77539-612
- 77539-616
- 77539-614
- Protein/Peptide Name:
- GNB1
- GNB1
- GNB1
- Protein Synonyms:
- Guanine Nucleotide Binding Protein (G Protein), Beta Polypeptide 1
- Guanine Nucleotide Binding Protein (G Protein), Beta Polypeptide 1
- Guanine Nucleotide Binding Protein (G Protein), Beta Polypeptide 1
- Protein/Peptide Type:
- Recombinant
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Sequence:
- MGSSHHHHHH SSGLVPRGSH MGSMSELDQL RQEAEQLKNQ IRDARKACAD ATLSQITNNI DPVGRIQMRT RRTLRGHLAK IYAMHWGTDS RLLVSASQDG KLIIWDSYTT NKVHAIPLRS SWVMTCAYAP SGNYVACGGL DNICSIYNLK TREGNVRVSR ELAGHTGYLS CCRFLDDNQI VTSSGDTTCA LWDIETGQQT TTFTGHTGDV MSLSLAPDTR LFVSGACDAS AKLWDVREGM CRQTFTGHES DINAICFFPN GNAFATGSDD ATCRLFDLRA DQELMTYSHD NIICGITSVS FSKSGRLLLA GYDDFNCNVW DALKADRAGV LAGHDNRVSC LGVTDDGMAV ATGSWDSFLK IWN
- MGSSHHHHHH SSGLVPRGSH MGSMSELDQL RQEAEQLKNQ IRDARKACAD ATLSQITNNI DPVGRIQMRT RRTLRGHLAK IYAMHWGTDS RLLVSASQDG KLIIWDSYTT NKVHAIPLRS SWVMTCAYAP SGNYVACGGL DNICSIYNLK TREGNVRVSR ELAGHTGYLS CCRFLDDNQI VTSSGDTTCA LWDIETGQQT TTFTGHTGDV MSLSLAPDTR LFVSGACDAS AKLWDVREGM CRQTFTGHES DINAICFFPN GNAFATGSDD ATCRLFDLRA DQELMTYSHD NIICGITSVS FSKSGRLLLA GYDDFNCNVW DALKADRAGV LAGHDNRVSC LGVTDDGMAV ATGSWDSFLK IWN
- MGSSHHHHHH SSGLVPRGSH MGSMSELDQL RQEAEQLKNQ IRDARKACAD ATLSQITNNI DPVGRIQMRT RRTLRGHLAK IYAMHWGTDS RLLVSASQDG KLIIWDSYTT NKVHAIPLRS SWVMTCAYAP SGNYVACGGL DNICSIYNLK TREGNVRVSR ELAGHTGYLS CCRFLDDNQI VTSSGDTTCA LWDIETGQQT TTFTGHTGDV MSLSLAPDTR LFVSGACDAS AKLWDVREGM CRQTFTGHES DINAICFFPN GNAFATGSDD ATCRLFDLRA DQELMTYSHD NIICGITSVS FSKSGRLLLA GYDDFNCNVW DALKADRAGV LAGHDNRVSC LGVTDDGMAV ATGSWDSFLK IWN
- Protease-free:
- No
- No
- No
- Animal-Free:
- No
- No
- No
- Nuclease-free:
- No
- No
- No
- Carrier-Free:
- No
- No
- No
- Endotoxin-free:
- No
- No
- No
- Endotoxin Level:
- Low
- Low
- Low
- UniProtKB:
- P62873
- P62873
- P62873
- Purity:
- >85% by SDS-PAGE
- >85% by SDS-PAGE
- >85% by SDS-PAGE
- Grade:
- Ultrapure grade
- Ultrapure grade
- Ultrapure grade
- Concentration:
- 0.25 mg/ml (Determined by Bradford assay)
- 0.25 mg/ml (Determined by Bradford assay)
- 0.25 mg/ml (Determined by Bradford assay)
- Molecular Weight:
- 39.8 kDa (363 aa)
- 39.8 kDa (363 aa)
- 39.8 kDa (363 aa)
- Storage Conditions:
- 4 °C
- 4 °C
- 4 °C
- Shipping Temperature:
- 20
- 20
- 20
Specifications
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: GNB1-172H-10UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77539-612Protein/Peptide NameGNB1Protein SynonymsGuanine Nucleotide Binding Protein (G Protein), Beta Polypeptide 1Protein/Peptide TypeRecombinantSpeciesHumanSourceE. coliConjugationUnconjugatedSequenceMGSSHHHHHH SSGLVPRGSH MGSMSELDQL RQEAEQLKNQ IRDARKACAD ATLSQITNNI DPVGRIQMRT RRTLRGHLAK IYAMHWGTDS RLLVSASQDG KLIIWDSYTT NKVHAIPLRS SWVMTCAYAP SGNYVACGGL DNICSIYNLK TREGNVRVSR ELAGHTGYLS CCRFLDDNQI VTSSGDTTCA LWDIETGQQT TTFTGHTGDV MSLSLAPDTR LFVSGACDAS AKLWDVREGM CRQTFTGHES DINAICFFPN GNAFATGSDD ATCRLFDLRA DQELMTYSHD NIICGITSVS FSKSGRLLLA GYDDFNCNVW DALKADRAGV LAGHDNRVSC LGVTDDGMAV ATGSWDSFLK IWNProtease-freeNoAnimal-FreeNoNuclease-freeNoCarrier-FreeNoEndotoxin-freeNoEndotoxin LevelLowUniProtKBP62873Purity>85% by SDS-PAGEGradeUltrapure gradeConcentration0.25 mg/ml (Determined by Bradford assay)Molecular Weight39.8 kDa (363 aa)Storage Conditions4 °CShipping Temperature20 - Supplier Number: GNB1-172H-50UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77539-616Protein/Peptide NameGNB1Protein SynonymsGuanine Nucleotide Binding Protein (G Protein), Beta Polypeptide 1Protein/Peptide TypeRecombinantSpeciesHumanSourceE. coliConjugationUnconjugatedSequenceMGSSHHHHHH SSGLVPRGSH MGSMSELDQL RQEAEQLKNQ IRDARKACAD ATLSQITNNI DPVGRIQMRT RRTLRGHLAK IYAMHWGTDS RLLVSASQDG KLIIWDSYTT NKVHAIPLRS SWVMTCAYAP SGNYVACGGL DNICSIYNLK TREGNVRVSR ELAGHTGYLS CCRFLDDNQI VTSSGDTTCA LWDIETGQQT TTFTGHTGDV MSLSLAPDTR LFVSGACDAS AKLWDVREGM CRQTFTGHES DINAICFFPN GNAFATGSDD ATCRLFDLRA DQELMTYSHD NIICGITSVS FSKSGRLLLA GYDDFNCNVW DALKADRAGV LAGHDNRVSC LGVTDDGMAV ATGSWDSFLK IWNProtease-freeNoAnimal-FreeNoNuclease-freeNoCarrier-FreeNoEndotoxin-freeNoEndotoxin LevelLowUniProtKBP62873Purity>85% by SDS-PAGEGradeUltrapure gradeConcentration0.25 mg/ml (Determined by Bradford assay)Molecular Weight39.8 kDa (363 aa)Storage Conditions4 °CShipping Temperature20 - Supplier Number: GNB1-172H-250UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77539-614Protein/Peptide NameGNB1Protein SynonymsGuanine Nucleotide Binding Protein (G Protein), Beta Polypeptide 1Protein/Peptide TypeRecombinantSpeciesHumanSourceE. coliConjugationUnconjugatedSequenceMGSSHHHHHH SSGLVPRGSH MGSMSELDQL RQEAEQLKNQ IRDARKACAD ATLSQITNNI DPVGRIQMRT RRTLRGHLAK IYAMHWGTDS RLLVSASQDG KLIIWDSYTT NKVHAIPLRS SWVMTCAYAP SGNYVACGGL DNICSIYNLK TREGNVRVSR ELAGHTGYLS CCRFLDDNQI VTSSGDTTCA LWDIETGQQT TTFTGHTGDV MSLSLAPDTR LFVSGACDAS AKLWDVREGM CRQTFTGHES DINAICFFPN GNAFATGSDD ATCRLFDLRA DQELMTYSHD NIICGITSVS FSKSGRLLLA GYDDFNCNVW DALKADRAGV LAGHDNRVSC LGVTDDGMAV ATGSWDSFLK IWNProtease-freeNoAnimal-FreeNoNuclease-freeNoCarrier-FreeNoEndotoxin-freeNoEndotoxin LevelLowUniProtKBP62873Purity>85% by SDS-PAGEGradeUltrapure gradeConcentration0.25 mg/ml (Determined by Bradford assay)Molecular Weight39.8 kDa (363 aa)Storage Conditions4 °CShipping Temperature20
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



