Order Entry
United States
ContactUsLinkComponent
Human Recombinant GNAQ (from HEK293T cells)
Human Recombinant GNAQ (from HEK293T cells)
Supplier:  Creative Biomart
New Product
Certificates
undefined
Human Recombinant GNAQ (from HEK293T cells)
Supplier:  Creative Biomart
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293T cells
  • Species:
    Human
  • Storage Conditions:
    4 °C
  • Endotoxin-free:
    No
  • Carrier-Free:
    No
  • Protease-free:
    No
  • Animal-Free:
    No
  • Protein Synonyms:
    guanine nucleotide binding protein (G protein), q polypeptide
  • UniProtKB:
    P50148
  • Protein/Peptide Name:
    GNAQ
  • Purity:
    >80% as determined by SDS-PAGE and coomassie blue staining
  • Molecular Weight:
    42 kDa
  • Endotoxin Level:
    Low
  • Concentration:
    >50 μg/ml as determined by microplate BCA method
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping Temperature:
    20

Specifications

Frequently Bought Together