Order Entry
United States
ContactUsLinkComponent
Human Recombinant FLT1 (from E. coli)
Human Recombinant FLT1 (from E. coli)
Supplier:  Creative Biomart
New Product
Certificates
undefined
Human Recombinant FLT1 (from E. coli)
Supplier:  Creative Biomart
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    4 °C
  • Endotoxin-free:
    No
  • Carrier-Free:
    No
  • Protease-free:
    No
  • Animal-Free:
    No
  • Protein Synonyms:
    fms-related tyrosine kinase 1
  • UniProtKB:
    P17948
  • Protein/Peptide Name:
    FLT1
  • Purity:
    >95% by SDS-PAGE
  • Sequence:
    SLKGTQHIMQAGQTLHLQCRGEAAHKWSLPEMVSKESERLSITKSACGRNGKQFCSTLTLNTAQANHTGFYSCKYLAVPTSKKKETESAIYIFISDTGRPFVEMYSEIPEIIHMTEGRELVIPCRVTSPNITVTLKKFPLDTLIPDGKRIIWDSRKGFIISNATYKEIGLLTCEATVNGHLYKTNYLTHRQTNTIIDVQISTPRPVKLLRGHTLVLNCTATTPLNTRVQMTWSYPDEKNKRASVRRRIDQSNSHANIFYSVLTIDKMQNKDKGLYTCRVRSGPSFKSVNTSVHI
  • Endotoxin Level:
    Low
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping Temperature:
    20
  • Tested Applications:
    Western blot, immunohistochemistry, ELISA, dot blot, SDS-PAGE

Specifications

Frequently Bought Together