Chicken Recombinant FGF7 (from E. coli)
Catalog # 77539-486
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Chicken
- Size:50 µg
- Storage Conditions:4 °C
- Endotoxin Content:<1 EU/μg by LAL
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:Fibroblast growth factor 7
- UniProtKB:Q5KRA4
- Protein/Peptide Name:FGF7
- Purity:>95% by SDS-PAGE
- Molecular Weight:The protein has a calculated MW of 21 kDa
- Sequence:MGSSHHHHHHSSGLVPRGSHMCNDMTPEQMATNVNCSSPERHTRSYDYMEGGDVRVRRLFCRTQWYMRIDKRGKVKGTREANNNYSILEIRTVAVGIVAIKGVESEYFLAMNKSGRLYGKKVCNEDCNFIELIEENHYNTYASAKWTHKGKEMFVTLNHKGVPMKGKKTKKEHRASHFLPLAIS
- Endotoxin Level:Low
- Concentration:67 μg/ml by BCA
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Cat. No.:77539-486