Human Recombinant DYNLL1 (from E. coli)
77539-324
New Product
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:100 µg
- Storage Conditions:4 °C
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:dynein, light chain, LC8-type 1
- UniProtKB:P63167
- Protein/Peptide Name:DYNLL1
- Purity:>90% by SDS-PAGE
- Endotoxin Level:Low
- Concentration:1 mg/ml (Determined by Bradford assay)
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Cat. No.:77539-324