Human Recombinant CD200R1 (from Wheat germ)
Catalog # 77538-880
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Wheat germ
- Species:Human
- Size:2 µg
- Storage Conditions:4 °C
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:CD200 receptor 1
- UniProtKB:Q8TD46
- Protein/Peptide Name:CD200R1
- Molecular Weight:65.4 kDa
- Sequence:MLCPWRTANLGLLLILTIFLVAEAEGAAQPNNSLMLQTSKENHALASSSLCMDEKQITQNYSKVLAEVNTSWPVKMATNAVLCCPPIALRNLIIITWEIILRGQPSCTKAYKKETNETKETNCTDERITWVSRPDQNSDLQIRTVAITHDGYYRCIMVTPDGNFHRGYHLQVLVTPEVTLFQNRNRTAVCKAVAGKPAAHISWIPEGDCATKQEYWSNGTVTVKSTCHWEVHNVSTVTCHVSHLTGNKSLYIELLPVPGAKKSAKLYIPYIILTIIILTIVGFIWLLKVNGCRKYKLNKTESTPVVEEDEMQPYASYTEKNNPLYDTTNKVKASEALQSEVDTDLHTL
- Endotoxin Level:Low
- Concentration:0.03 µg/µl
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Tested Applications:Array, AP, ELISA, WB-Re
- Cat. No.:77538-880